missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Profilin 2 Polyclonal specifically detects Profilin 2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | Profilin 2 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | D3S1319E, PFL, profilin 2, Profilin II, profilin-2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Profilin 2 (AAH18049.1; Met1-Phe140). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?