missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Profilin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527612
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, mouse spleen, Rat spleen. Predicted reactivity: Mouse (95%), Rat (95%), Pig (94%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Profilin is an ubiquitous small (12-15 kDa) phosphoinositides and poly-L-proline binding protein that plays a role in both signal transduction pathways and actin filament dynamics. There are two mammalian profilins with similar biochemical properties but different expression patterns. Profilin I appears to be highly expressed in most tissues except for skeletal muscle. Profilin II is predominantly expressed in brain and at lower levels in skeletal muscle, uterus and kidney. Profilin is a mainly cytosolic protein with higher concentrations in dynamic membrane areas like the leading edge and ruffling membranes.
Specifications
| Profilin 1 | |
| Polyclonal | |
| Unconjugated | |
| Pfn1 | |
| actin binding protein; ALS18; Epididymis tissue protein Li 184a; Pfn; Pfn1; profilin 1; profilin I; Profilin-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 18643, 5216, 64303 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.56 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P07737, P62962, P62963 | |
| Pfn1 | |
| Recombinant protein encompassing a sequence within the center region of human Profilin 1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction