missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Profilin 1 Antibody (S06-6H7), Novus Biologicals™
Rabbit Monoclonal Antibody
£328.00 - £500.00
Specifications
| Antigen | Profilin 1 |
|---|---|
| Clone | S06-6H7 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, ELISA, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin, Sandwich ELISA |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18616604
|
Novus Biologicals
NBP3-19483-50ul |
50 μg |
£328.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625364
|
Novus Biologicals
NBP3-19483-100ul |
100 μg |
£500.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Profilin 1 Monoclonal antibody specifically detects Profilin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISASpecifications
| Profilin 1 | |
| 0.3 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Sandwich ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 5216 | |
| IgG | |
| Affinity purified |
| S06-6H7 | |
| Western Blot, ELISA, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin, Sandwich ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| profilin 1, Profilin I, profilin-1 | |
| A synthetic peptide of human Profilin 1 (Uniprot # P07737) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title