missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Profilin 1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-19344UV
This item is not returnable.
View return policy
Description
Profilin 1 Polyclonal specifically detects Profilin 1 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Profilin 1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| profilin 1, Profilin I, profilin-1 | |
| Rabbit | |
| Affinity Purified | |
| Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
| 5216.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PFN1 | |
| Synthetic peptide made to a C-terminal portion of human Profilin 1 (within residues 100-140). [Swiss-ProtNo. P07737] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction