Learn More
Profilin 1 Antibody (CL3524) [DyLight 550], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59778R
Product Code. 30301398
  • £558.00 / 0.10mL
Description
Profilin 1 Monoclonal antibody specifically detects Profilin 1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out,KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Profilin 1
Monoclonal
DyLight 550
profilin 1, Profilin I, profilin-1
This antibody was developed using a recombinant protein derived from P07737, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Tumor Suppressors
5216
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out, KnockDown
CL3524
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified