missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Profile Sensa XP™ Latex Gloves

Product Code. 12705785
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 12705785

missing translation for 'mfr': Nitritex™ PSXP202M

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High strength and excellent sensitivity. Nitritex™ Profile Sensa XP™ Latex Gloves are extra-length, powder-free examination gloves that measure 300mm/12 in., providing additional coverage past the wrist. Certified to Class I Medical and Cat 3 PPE standards.

  • Natural rubber latex provides exceptional strength while still providing excellent sensitivity
  • Mid-arm ambidextrous gloves measure 300mm for more coverage
  • Beaded cuff for easier donning and stability on the arm
  • Provides improved grip with a textured surface
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 455, EN 374; AQL 1,5
Classification Category III PPE, Class I Medical
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Hand Ambidextrous
Material Latex
Powdered Powder Free
Size Medium
Sterility Non-sterile
Glove Cuff Style Beaded
Packaging Type Dispenser box
Quantity 100 Pk.
Length (English) Cuff 12 in.
Length (Metric) Cuff 300 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.