missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Profile Gel™ Sterile Latex Surgical Gloves

Product Code. 15591671
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
This item is not returnable. View return policy

Product Code. 15591671

Brand: Nitritex™ PGHS70

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Sterile Latex Surgical Gloves are powder free, hand-specific gloves with a polymer coating for improved softness. Beaded cuff is 50% thicker cuff to prevent tearing and textured surface helps to ensure secure grip.

  • Latex for strength and comfort
  • Extra thick cuffs for strength
  • Inner polymer gel coating enables easy smooth donning
  • Textured finish
  • Anatomically shaped to reduce wearer fatigue
  • Powder free
  • Class IIa Medical
  • Complies with the requirements of the European Medical Gloves Standard EN 455 (Parts 1, 2 & 3)
  • Manufactured in a facility holding ISO 9001:2008 certification
  • Hand specific
  • 300 mm length
  • Sterile
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Complies with the requirements of the European Medical Gloves Standard EN 455 (Parts 1, 2 & 3)
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Hand Specific
Latex Free No
Material Latex
Powdered Powder Free
Size 7
Sterility Sterile
Packaging Type EasyTear™ pouches in a box
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.