missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Profile Gel™ Sterile Latex Surgical Gloves

Catalog Number 15541671
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
This item is not returnable. View return policy
This item is not returnable. View return policy

Sterile Latex Surgical Gloves are powder free, hand-specific gloves with a polymer coating for improved softness. Beaded cuff is 50% thicker cuff to prevent tearing and textured surface helps to ensure secure grip.

  • Latex for strength and comfort
  • Extra thick cuffs for strength
  • Inner polymer gel coating enables easy smooth donning
  • Textured finish
  • Anatomically shaped to reduce wearer fatigue
  • Powder free
  • Class IIa Medical
  • Complies with the requirements of the European Medical Gloves Standard EN 455 (Parts 1, 2 & 3)
  • Manufactured in a facility holding ISO 9001:2008 certification
  • Hand specific
  • 300 mm length
  • Sterile
Certifications/Compliance Complies with the requirements of the European Medical Gloves Standard EN 455 (Parts 1, 2 & 3)
Color Natural
Disposable Single-use
Exterior Finish Fully Textured
Hand Specific
Latex Free No
Material Latex
Powdered Powder Free
Size 6.5
Sterility Sterile
Packaging Type EasyTear™ pouches in a box
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'