missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Omega Pro-tech XP™ Nitrile Gloves

Product Code. 15175544
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Each
Pack of 100
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15175544 Large Each
13455259 Medium Pack of 1000
12735875 X-Large Pack of 100
14891292 Small Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15175544 Supplier Nitritex™ Supplier No. OPTXP2104L
  • ONLINE EXCLUSIVE PRICE
    £301.40
    £200.12 / Pack of 1
    Save £101.28
    33% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

The premier lab glove delivers excellent protection and exceptional tactility. Nitritex™ Omega Pro-tech XP™ Nitrile Gloves are non-sterile, extra-length laboratory gloves that are 100% latex free, thereby eliminating all risk of Type 1 latex allergic reactions.

  • Violet nitrile material is resistant to a wide range of chemicals, fatty acids and bodily fluids
  • Ambidextrous gloves are 300mm (12 in.) in length to provide extra protection in the cuff area
  • Strong puncture resistance with no compromise on tactile performance and dexterity
  • Beaded cuff for easier donning and stability on the arm
  • Texture surface delivers improved grip
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN420:2003 & EN374-1 & 2:2003
Classification European PPE Category 3 Complex Design
Color Purple
Disposable Single-use
Exterior Finish Partially Textured
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Large
Sterility Non-sterile
Glove Cuff Style Beaded
Packaging Type Dispenser Box
Resistant To Chemicals, bodily fluids, fatty acids
Length (English) Cuff 12 in.
Length (Metric) Cuff 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.