Learn More
The Daylight Company Ltd Professional Table Base
Click to view available options
Color:
White
Description
- Heavy-duty table base designed to support the weight of Daylight lamps and magnifiers.
- Compatible with the following products: Omega 5 Magnifier (D25110), Omega 7 Magnifier (D25130), Lumi (D35500), iQ Magnifier (25100), MAG Lamp S Magnifier (N1200), MAG Lamp XL Magnifier (N1300)
Specifications
Specifications
| Product Type | Stand Base |
| Autoclavable | Not Autoclavable |
| Color | White |