missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prodynorphin Recombinant Rabbit Monoclonal Antibody (HL2574)

Description
Prodynorphin Recombinant Monoclonal Antibody for IHC (P), Western Blot
expression in hippocampus increases in response to traumatic brain injury; may play a role in regulatio of neurotransmitter release.
Specifications
Specifications
| Antigen | Prodynorphin |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | HL2574 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Pdyn |
| Gene Accession No. | P01213 |
| Gene Alias | ADCA; Alpha-neoendorphin; beta-neoendorphin; Beta-neoendorphin-dynorphin; Big Dyn; Big dynorphin; Dyn; Dyn-A17; Dyn-B; Dynorphin A(1-13); dynorphin A(1-17); Dynorphin A(1-8); Dynorphin B; Dynorphin B(1-13); Dynorphin B-29; Leu-enkephalin; leumorphin; neoendorphin-dynorphin-enkephalin prepropeptide; PDYN; PENKB; Preprodynorphin; preproenkephalin B; prodynorphin; proenkephalin B; proenkephalin-B; Rimorphin; SCA23; unnamed protein product |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?