missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prodynorphin Recombinant Rabbit Monoclonal Antibody (HL2573)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556844
This item is not returnable.
View return policy
Description
Prodynorphin Recombinant Monoclonal Antibody for Western Blot
expression in hippocampus increases in response to traumatic brain injury; may play a role in regulatio of neurotransmitter release.
Specifications
| Prodynorphin | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P01213 | |
| Pdyn | |
| Recombinant fragment of human ProDynorphin. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL2573 | |
| Unconjugated | |
| Pdyn | |
| ADCA; Alpha-neoendorphin; beta-neoendorphin; Beta-neoendorphin-dynorphin; Big Dyn; Big dynorphin; Dyn; Dyn-A17; Dyn-B; Dynorphin A(1-13); dynorphin A(1-17); Dynorphin A(1-8); Dynorphin B; Dynorphin B(1-13); Dynorphin B-29; Leu-enkephalin; leumorphin; neoendorphin-dynorphin-enkephalin prepropeptide; PDYN; PENKB; Preprodynorphin; preproenkephalin B; prodynorphin; proenkephalin B; proenkephalin-B; Rimorphin; SCA23; unnamed protein product | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5173 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction