missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prodynorphin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115150
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Dynorphin A.
expression in hippocampus increases in response to traumatic brain injury; may play a role in regulatio of neurotransmitter release.
Specifications
| Prodynorphin | |
| Polyclonal | |
| Unconjugated | |
| Pdyn | |
| ADCA; Alpha-neoendorphin; beta-neoendorphin; Beta-neoendorphin-dynorphin; Big Dyn; Big dynorphin; Dyn; Dyn-A17; Dyn-B; Dynorphin A(1-13); dynorphin A(1-17); Dynorphin A(1-8); Dynorphin B; Dynorphin B(1-13); Dynorphin B-29; Leu-enkephalin; leumorphin; neoendorphin-dynorphin-enkephalin prepropeptide; PDYN; PENKB; Preprodynorphin; preproenkephalin B; prodynorphin; proenkephalin B; proenkephalin-B; Rimorphin; SCA23; unnamed protein product | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18610, 29190, 5173 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O35417, P01213, P06300 | |
| Pdyn | |
| A synthesized peptide derived from human PDYN(Accession P01213), corresponding to amino acid residues G195-N245. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction