missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Microwave Stirrer

Product Code. 12348229 Shop All SP Scienceware Products
Change view
Click to view available options
Dimensions (L x W x H):
276 x 257 x 67 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
12348229 276 x 257 x 67 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12348229 Supplier Bel-Art™ SP Scienceware™ Supplier No. F370400000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Converts the motion from the turntable impeller inside a microwave oven into a magnetic stirrer allowing simultaneous heating and stirring for increased efficiency

  • Heated stirring can improve dissolving and mixing; permits preparation of super-saturated solutions
  • Ideal for melting agar and agarose, concentrating urea solutions, or other applications that benefit from heated stirring, and for chemical purification
  • Adjustable pins make stirrer compatible with most microwave drive spindles
  • Can be used with magnetic stirring bars up to 5cm in length

Specifications

Dimensions (L x W x H) 276 x 257 x 67 mm
Height (Metric) 67 mm
Length (Metric) 276 mm
Width (Metric) 257 mm
Color White
Type Stirrer Microwave
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.