Learn More
Thermo Scientific™ Productainer™ BioProcess Container (BPC), 100 L with 4 TopPorts and 1 Bottom Dispense, 3D Single-Use BPC

The Thermo Scientific™ Productainer™ BioProcess Container (BPC), 100 L 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 4 top ports and 1 bottom port. The 3D square tube design with bottom port dispense is for use with conical, bottom-drain drums.
Brand: Thermo Scientific™ SH30652.02
Description
Features:
- Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
- All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
- All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000 L.
Applications of the Thermo Scientific 3D BPC include:
- Hydration and sterilization of process buffers, liquids, and culture media.
- Chromatography feed and fraction collection.
- Storage and transport of bulk drug product and bulk drug precursors.
- Harvest from and feed into bioreactors and fermentors.
- Dispensing, packaging, and storage of cell culture media, buffers and process liquids.
Customize standard BPC systems for optimized single-use technology. Choose from the industry’s largest component library – with over 20,000 unique components that include:
- Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
- Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
- Filters—used for bioburden reduction; sterilization grade filters and vent filters.
- Tubing—thermoplastic elastomers, platinum-cured silicone.
- Sample ports—septum-style and needle-free ports.
- Clamps—sliders, pinch, and crimp bands.
Specifications
| 100 L | |
| Bottom Port: 1 x 0.38 in., Top Ports: 1 x 0.38 in. 2 x 0.25 in. | |
| Packages placed in strong cardboard cartons with product and lot identification | |
| Gamma Irradiated | |
| Line 2. Insert luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 in. x 0.38 in.) Top Port | |
| Line 5. MPC body, polycarbonate Tubing: silicone, length: 121.9 cm (48 in.) ID x OD: 9.5 x 15.9 mm (0.38 in. x 0.63 in.) Bottom port | |
| Productainer™ |
| CX5-14 | |
| SV50517.05 | |
| Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area | |
| Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 121.9 cm (48 in.) dip tube ID x OD: 9.5 x 15.9 mm (0.38 in. x 0.63 in.) with dip tube length: 30.5 cm (12 in.) Top Port | |
| Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 in. x 0.38 in.) Top Port | |
| Line 4. Plugged, no tubing |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.