Learn More
Thermo Scientific™ Productainer™ 3D BioProcess Container (BPC), Aegis™ 5-14 film, five ports, Luer locks + MPC
Shop All Thermo Scientific Products

Description
Productainer BPCs are constructed with Aegis 5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainer BPCs are suitable for a wide array of bioprocess applications. For optimal fit, form, and function, Productainer BPCs can be customized using the industry's largest component library.
Highlights:
• Film optimized for flexibility and durability—superior stress resistance, puncture resistance, and physical barrier properties to ensure stability of contents
• Gamma-irradiation sterilized
• Squared tube design with polyethylene ports located on the top of the BPC for easy access
• Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
• Customizable via the industry's largest library of qualified components
Recommended for:
• Harvest—collection and storage of bulk product from bioreactors
• Storage—sterile storage of media, buffers, and process intermediates
• Separation—feeding and receiving of sterile liquids from separation systems
• Purification—buffer feeding to purification system, fraction collection and storage
• Filling—sterile reservoir for filling systems
• Transport and shipping—bulk sterile liquids, buffers, media and process intermediates within and between facilities
Specifications
Specifications
| Capacity (Metric) | 200 L |
| Description | Bottom Drain |
| Film Description | Aegis 5-14 |
| Port Detail | 5 ports |
| Quantity | 200 L |
| Tubing Style | C-Flex |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.