missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Productainer™ 3D BioProcess Container, BioTitan retainers, four ports, MPX and Luer locks

Product Code. 31940836 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
Unit Size:
Pack of 10
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
31940836 50 L Pack of 5
17958271 50 L Pack of 10
17918241 100 L Pack of 5
17948271 200 L Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 31940836 Supplier Thermo Scientific™ Supplier No. SH31299.01
 Restricted Item
Estimated Shipment: 25-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use Thermo Scientific Productainer BPC bags with BioTitan retainers to package, store, and transport media, buffers, water for injection (WFI), process liquids, and biological products.

Productainer BPCs are constructed with our Thermo Scientific Aegis5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility. Our standard Productainers are suitable for a wide array of bioprocess applications. For optimal fit, form and function, Productainers can be customized using the industry's largest component library.

Features include:

  • Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
  • Gamma-irradiation sterilized
  • Squared tube design with polyethylene ports located on the top of the BPC for easy access
  • Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
  • Customizable via the industry's largest library of qualified components
  • Now manufactured with the BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points

Recommended for:

  • Harvest—collection and storage of bulk product from bioreactors
  • Storage—sterile storage of media, buffers and process intermediates
  • Separation—feeding and receiving of sterile liquids from separation systems
  • Purification—buffer feeding to purification system, fraction collection and storage
  • Filling—sterile reservoir for filling systems
  • Transport and Shipping—bulk sterile liquids, buffers, media and process intermediates within and between facilities
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 50 L
For Use With (Application) Single-Use Bioprocessing
Quantity 50 L
Tubing Style TPE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.