missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ E-Gel™ Power Snap Electrophoresis Device Starter Kit, SYBR Safe 1.2%
Click to view available options
Max. Voltage:
240 V AC (Power Supply), 48 V DC (Electrophoresis Device)
missing translation for 'unitSize'
Each
Description
Includes
E-Gel Power Snap Electrophoresis device,orange viewing glasses,E-Gel Go! agarose gel adaptor,region specific cable with power adaptor.
The Invitrogen E-Gel Power Snap Electrophoresis Device Starter Kit, SYBR Safe 1.2%, provides everything you need to quickly start using precast 1.2% E-Gel agarose gels with SYBR Safe on the device.
The kit includes:
- 18 E-Gel agarose gels with SYBR Safe, 1.2% (Cat. No. G521801)
- E-Gel Power Snap Electrophoresis Device (Cat. No. G8100)
- E-Gel 1 Kb Plus DNA Ladder with E-Gel Sample Loading Buffer (1X) (Cat. No. 10488090)
Services
1 year warranty
Specifications
Specifications
| Content And Storage | • 1 electrophoresis device • 18 E-Gel SYBR Safe 1.2% gels • 1 vial E-Gel 1 kb Plus DNA Ladder • 1 power adapter • 1 pair of viewing glasses • 1 E-Gel Go! Adapter Store at room temperature |
| Type | Electrophoresis Device Starter Kit |
| Shipping Condition | Room Temperature |
| Max. Voltage | 240 V AC (Power Supply), 48 V DC (Electrophoresis Device) |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
