missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ProFlex™ PCR System, 2 x Flat

The Veriti 96-well fast thermal cycler delivers the proven reliability of Applied Biosystems instruments. With the added control of VeriFlex blocks, you have six independent temperature blocks that provide precise control over your PCR optimization.

Brand:  Applied Biosystems™ 4484078


6627.50 GBP valid until 2024-06-28
Use promo code "21649" to get your promotional price.



Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15182099

  • £12050.00 / Each
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Veriti 96-well fast thermal cycler delivers the proven reliability of Applied Biosystems instruments. With the added control of VeriFlex blocks, you have six independent temperature blocks that provide precise control over your PCR optimization. Additionally, the color touchscreen simplifies the setup and use of the Veriti 96-well fast thermal cycler. Optional setups for fast or standard PCR methods provide flexibility to shorten your PCR cycling times. The Veriti 96-well fast thermal cycler comes with a 0.1-mL block configuration that saves time and reagent consumption.

Features of the Veriti 96-well fast thermal cycler include:
• Innovative VeriFlex blocks allow for precise PCR optimization or to run up to six independent assays
• Six temperature zones for convenient PCR optimization
• Standard- and fast-enabled cycling to address current and future PCR needs
• Easy-to-use 6.5-inch VGA touchscreen
• Convenient protocol transfer from one Veriti thermal cycler to another using USB
• Reduced PCR cycling time when using faster ramp rates
• Optional VeritiLink Remote Management Software allows remote management of more than 50 cyclers

VeriFlex blocks enhanced PCR functionality
Separate peltier blocks provide maximal versatility, flexibility, and offer two key benefits:
1. Optimization—each block can be set with a specific temperature for precise control over PCR optimization
2. More experiments—six peltier blocks can be used to run different annealing temperatures in the same run

Control at your fingertips
The powerful, yet simple to operate user interface on the Veriti system is driven by the 6.5-inch (16.51-cm) VGA color touchscreen. The large screen allows for easy viewing of temperature profiles, and large navigation buttons put programming at your fingertips. Setup and navigation of the Veriti Thermal Cycler does not require the use of a stylus or mouse.

Fast setup, fast results

The Veriti system has two options for method navigation. For quick setup, you may select one of the pre-programmed methods to run. If you prefer to program your own methods, simply touch the step you would like to setup and then use the keypad to enter thermal cycling steps. After completing a setup, you have the choice of saving the method to the instrument or to a USB memory stick.

Control your time, go fast when you want

With the Veriti Thermal Cycler you get two thermal cyclers in one: a standard thermal cycler with its better than gradient feature and a fast-enabled thermal cycler. So whether you have a current need to run fast PCR and shorten your PCR cycling times, or a possible future application need for fast PCR, the Veriti Thermal Cycler provides you with the flexibility to go fast when you want.

Applied Biosystems PCR plastics
Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment. To find the best match please visit our Plastics Selection Guide.

Specifications

Specifications

Interchangeable
High-throughput Compatible
Fast, Standard
8 OpenArray Plates, 24 x 20K Chips
8.4 in. Color TFT LCD Touchscreen
Limited Warranty
PCR
USB, WiFi, Cloud, Ethernet
USB and On-board
2 x Flat Blocks
±0.25°C (35°C to 99.9°C)
<0.5°C (20 sec. after reaching 95°C)
22 x 13 x 10.6 in. (56.5 x 33 x 27 cm)
100/240 V
Thermal Cycler
0°C to 100°C
Auto re-start (after Power Outages), Program Overwrite Protection
3°C/sec. (Block), 1.6°C/sec. (Sample)
45 lb. (20.4 kg)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.