missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Bolt™ Western Pack B (PVDF)

Product Code. 15145204
Click to view available options
Dimensions (LxW):
Mini, 8 cm x 8 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15145204

Brand: Invitrogen™ B1000B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

2 Mini Blot Modules, 125mL Bolt™ Transfer Buffer (20X), 15mL Bolt™ Antioxidant, 20-pack PVDF membranes (0.2μm)

A wet transfer device used exclusively with the Mini Gel Tank

The tank accommodates one blot module per chamber, or two blot modules total with the side-by-side layout. The Mini Blot Module is compatible with Bolt™ Bis-Tris Plus gels and accommodates one gel per wet transfer. The universal connection and molded gasket make the blot module easy to use, while the inner core of the blot module allows for use of less methanol-based transfer buffer per wet transfer. At the recommended conditions and constant voltage, proteins can be transferred to nitrocellulose or PVDF membranes typically in 30 to 60 minutes.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Bolt™
Quantity ≤ 2 Blot Modules/Mini-Gel Tank, 1 Mini-Gel/Blot Module kit
Material PVDF
Content And Storage 2 Mini Blot Modules
125 mL Bolt™ Transfer Buffer (20X)
15 mL Bolt™ Antioxidant
20-pack PVDF membranes (0.2 μm)
Mode of Transfer Wet
Dimensions (LxW) Mini, 8 cm x 8 cm
Product Line Bolt

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.