missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates cellGrade™ 384-Well, Flat-Bottom Microplate
Color:Black; Sterility:Sterile; Lid:With Lid
Brand: BRAND™ 781982
384.56 GBP valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Code : 03
Additional Details : Weight : 2.61000kg
Description
- Polystyrene construction
- Sterile
- Lids included
- Standard plate for the cultivation of adherent cell lines
- PS-surface with different chemical groups, such as carboxyl and hydroxyl groups, that are freely accessible
- Surface is hydrophilic compared with non-treated PS
- Serum components are easily bound onto the freely accessible chemical groups, allowing an indirect adhesion of cells
- Clearly distinguishable through the color code: orange, embossed alphanumeric coding for 96-well standard plates
- Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
Specifications
ANSI, SLAS | |
With Lid | |
Carboxyl and Hydroxyl Groups | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
Cultivation of adherent cell lines | |
Yes |
Standard | |
16 x 24 | |
Black | |
384 | |
100 μL | |
Yes | |
50 |