missing translation for 'onlineSavingsMsg'
Learn More

n-(1,3-benzodioxol-5-ylmethyl)-n-methylamine, 97%, Thermo Scientific™

Product Code. 10214744
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10214744 250 mg 250mg
10796144 1 g 1g
10726485 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10214744 Supplier Thermo Scientific Maybridge Supplier No. CC01646CB
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 15205-27-3
Molecular Formula C9H12NO2
Molecular Weight (g/mol) 166.20
MDL Number MFCD04496422
InChI Key CEPGPPSMCRKGFJ-UHFFFAOYSA-O
Synonym n-1,3-benzodioxol-5-ylmethyl-n-methylamine, 1-1,3-benzodioxol-5-yl-n-methylmethanamine, 2h-1,3-benzodioxol-5-ylmethyl methyl amine, n-methyl-3,4-methylenedioxy benzylamine, 1,3-benzodioxole-5-methanamine,n-methyl, 1,3-benzodioxole-5-methanamine, n-methyl, 2h-1,3-benzodioxol-5-yl methyl methyl amine, 1,3-benzodioxol-5-ylmethyl methylaminehydrochloride, n-methylpiperonylamine, 3,4-methylenedioxy-n-methylbenzylamine
PubChem CID 421238
SMILES C[NH2+]CC1=CC=C2OCOC2=C1

Specifications

Assay Percent Range 97%
Quantity 250 mg
IUPAC Name [(2H-1,3-benzodioxol-5-yl)methyl](methyl)azanium
Formula Weight 165.19
Percent Purity 97%
Chemical Name or Material N-(1, 3-Benzodioxol-5-ylmethyl)-N-methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.