missing translation for 'onlineSavingsMsg'
Learn More

(6-Fluoro-4H-1,3-benzodioxin-8-yl)methanol, 90%, Thermo Scientific™

Product Code. 10150822
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10150822 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10150822 Supplier Thermo Scientific Maybridge Supplier No. CC01909DA
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 306934-89-4
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Molecular Formula C9H9FO3
MDL Number MFCD02677744
Quantity 1 g
Synonym 6-fluoro-4h-1,3-benzodioxin-8-yl methanol, 6-fluoro-4h-benzo d 1,3 dioxin-8-yl methanol, 6-fluoro-2,4-dihydro-1,3-benzodioxin-8-yl methanol, 4h-1,3-benzodioxin-8-methanol,6-fluoro, 6-fluoranyl-4h-1,3-benzodioxin-8-yl methanol, 6-fluoro-8-hydroxymethyl-4h-1,3-benzodioxine, 6-fluoro-2h,4h-benzo e 1,3-dioxan-8-yl methan-1-ol
InChI Key YLADDVZIEBXWQU-UHFFFAOYSA-N
SMILES C1C2=CC(=CC(=C2OCO1)CO)F
IUPAC Name (6-fluoro-4H-1,3-benzodioxin-8-yl)methanol
Molecular Weight (g/mol) 184.166
PubChem CID 2779909
Formula Weight 184.17
Percent Purity 90%
Chemical Name or Material (6-Fluoro-4H-1, 3-benzodioxin-8-yl)methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.