missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Paclitaxel, 99+%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Unit Size:
1mg
25mg
5mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Paclitaxel is a naturally ocurring compound originally isolated from the tree T. brevifolia
- Paclitaxel binds tubulin and stabilizes microtubule formation, preventing disassembly
- Paclitaxel is used as a drug to arrest cells at the mitotic checkpoint and block cell cycle progression, thus inducing death of cancer cells
Applications
- Paclitaxel can be used in preclinical laboratory research on diseases such as cancer, heart disease, inflammatory disorders, and hepatic fibrosis
- Paclitaxel can be used as a tool in cell biology to study mitotic checkpoint arrest conditions and downstream signaling cascades
Specifications
Specifications
| Melting Point | 213.0°C to 216.0°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (HPLC) |
| Quantity | 25 mg |
| Merck Index | 15, 7081 |
| Solubility Information | Solubility in water: sparingly soluble. Other solubilities: soluble in methanol,dmso,acetonitrile |
| Formula Weight | 853.9 |
| Percent Purity | 99+% |
| Physical Form | Powder |
| Show More |
Safety and Handling
Product Identifier
- Paclitaxel
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H361-Suspected of damaging fertility or the unborn child.
- H413-May cause long lasting harmful effects to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction