Learn More
R&D Systems™ ProDots Recombinant Human FGF basic/FGF2 (146 aa) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Specifications
| Accession Number | PO9O38 |
|---|---|
| Molecular Weight (g/mol) | M.W. Observed: 17 kDa, reducing conditions |
| Name | ProDots™ Recombinant Human FGF basic |
| Reconstitution | For a stock solution, reconstitute at 50μg/mL in sterile PBS, or simply roll ProDot directly into cell culture medium for immediate use. |
| Source | E. coli-derived human FGF basic/FGF2/bFGF protein Pro143 - Ser288 |
Includes: Individual vials include one ball of ProDots protein containing 5μg of Recombinant Human FGF basic.
Description
The ProDots Recombinant Human FGF basic/FGF2 (146 aa) Protein is derived from E. coli. The ProDots Recombinant Human FGF basic/FGF2 (146 aa) Protein has been validated for the following applications: Bioactivity.Order Info
Add ProDots proteins directly to the medium or reconstitute in the vial at 50μg/mL in sterile PBS.
Specifications
| PO9O38 | |
| ProDots™ Recombinant Human FGF basic | |
| E. coli-derived human FGF basic/FGF2/bFGF protein Pro143 - Ser288 | |
| Yes |
| M.W. Observed: 17 kDa, reducing conditions | |
| For a stock solution, reconstitute at 50μg/mL in sterile PBS, or simply roll ProDot directly into cell culture medium for immediate use. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt at room temperature as supplied. 12 months from date of receipt at 2-8° C as supplied. 1 month at 2-8° C under sterile conditions after reconstitution. 3 months at -20 to -80° C under sterile conditions after reconstitution. |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.