missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ ProDots Human IGF-I Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Specifications
| Gene Alias | IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C |
|---|---|
| Source | E. coli-derived human IGF-I/IGF-1 protein Gly49-Ala118 |
| Conjugate | Unconjugated |
| Recombinant | Yes |
| Gene ID (Entrez) | 3479 |
Description
The ProDots Recombinant Human IGF-I/IGF-1 Protein is derived from E. coli. The ProDots Recombinant Human IGF-I/IGF-1 Protein has been validated for the following applications: Bioactivity.Specifications
| IBP1, IGF1, IGF-1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, Somatomedin A, Somatomedin C, Somatomedin-C | |
| Unconjugated | |
| 3479 | |
| IGF1 | |
| <0.10 EU per 1μg of the protein by the LAL method. |
| E. coli-derived human IGF-I/IGF-1 protein Gly49-Ala118 | |
| Yes | |
| Observed M.W.: 7kDa, reducing conditions, Theoretical M.W.: 7.6kDa | |
| Measured in a serum-free cell proliferation assay using MCF-7 human breast cancer cells. Karey, K.P. et al. (1988) Cancer Research 48:4083. The ED50 for this effect is typically 0.3-1.5ng/mL. | |
| P05019 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title