missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ ProDots Human Flt-3 Ligand Recombinant Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Specifications
| Gene Alias | FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine |
|---|---|
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Flt-3 Ligand protein Thr27-Pro185 |
| Conjugate | Unconjugated |
| Recombinant | Yes |
| Gene ID (Entrez) | 2323 |
Description
The ProDots Recombinant Human Flt-3 Ligand Protein is derived from Sf 21 (baculovirus). The ProDots Recombinant Human Flt-3 Ligand Protein has been validated for the following applications: Bioactivity.Specifications
| FL, FLG3L, Flt3 ligand, Flt3L, FLT3LG, fms-related tyrosine kinase 3 ligand, SL cytokine | |
| Unconjugated | |
| 2323 | |
| FLT3LG | |
| <0.10 EU per 1μg of the protein by the LAL method. |
| Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Flt-3 Ligand protein Thr27-Pro185 | |
| Yes | |
| Predicted MW: 17.5kDa Observed MW: 17-30kDa, reducing conditions | |
| Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with mouse Flt-3. The ED50 for this effect is typically 0.2-1ng/mL. | |
| AAA17999.1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title