missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prodipine hydrochloride, MedChemExpress
Description
Prodipine, a diphenyl-phosphonate derivative. The IC50s of Prodipine for purified and plasma Dipeptidyl peptidase IV (DPP IV) from the rabbit are 4.5 μM and 30 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | Prodipine hydrochloride |
| CAS | 31314-39-3 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C20H26ClN |
| Quantity | 1 mg |
| SMILES | CC(C)N1CCC(C2=CC=CC=C2)(CC1)C3=CC=CC=C3.Cl |
| Molecular Weight (g/mol) | 315.88 |
| Formula Weight | 315.88 |
| Percent Purity | 97.29% |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?