missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRODH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527706
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (84%), Bovine (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a mitochondrial proline dehydrogenase that catalyzes the first step in proline degradation. Defects in this gene are a cause of hyperprolinemia type 1 and possibly susceptibility to schizophrenia 4 (SCZD4). The gene is located on chromosome 22q11.21, a region which has also been associated with the contiguous gene deletion syndromes DiGeorge syndrome and CATCH22 syndrome.
Specifications
| PRODH | |
| Polyclonal | |
| Unconjugated | |
| Prodh | |
| HSPOX2; p53-induced gene 6 protein; PIG6; POX; POX2; Pro1; Pro-1; Prodh; PRODH 1; PRODH 2; PRODH1; PRODH2; proline dehydrogenase; proline dehydrogenase (oxidase) 1; proline dehydrogenase (proline oxidase); proline dehydrogenase 1; proline dehydrogenase 1, mitochondrial; proline dehydrogenase, mitochondrial; proline oxidase; Proline oxidase 2; proline oxidase, mitochondrial; SCZD4; TP53I6; tumor protein p53 inducible protein 6; Ym24d07 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5625 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| O43272 | |
| Prodh | |
| Recombinant protein encompassing a sequence within the center region of human PRODH. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction