missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRODH Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142811
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize both reported isoforms (NP_057419.4; NP_001182155.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
The protein encoded by this gene is a mitochondrial proline dehydrogenase that catalyzes the first step in proline degradation. Defects in this gene are a cause of hyperprolinemia type 1 and possibly susceptibility to schizophrenia 4 (SCZD4). The gene is located on chromosome 22q11.21, a region which has also been associated with the contiguous gene deletion syndromes DiGeorge syndrome and CATCH22 syndrome.
Specifications
| PRODH | |
| Polyclonal | |
| Unconjugated | |
| Prodh | |
| HSPOX2; p53-induced gene 6 protein; PIG6; POX; POX2; Pro1; Pro-1; Prodh; PRODH 1; PRODH 2; PRODH1; PRODH2; proline dehydrogenase; proline dehydrogenase (oxidase) 1; proline dehydrogenase (proline oxidase); proline dehydrogenase 1; proline dehydrogenase 1, mitochondrial; proline dehydrogenase, mitochondrial; proline oxidase; Proline oxidase 2; proline oxidase, mitochondrial; SCZD4; TP53I6; tumor protein p53 inducible protein 6; Ym24d07 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5625 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| O43272 | |
| Prodh | |
| Peptide with sequence C-EDQESIQPLLRHYR (aa 121-134). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction