missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRODH Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | PRODH |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232650
|
Novus Biologicals
NBP3-38198-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231323
|
Novus Biologicals
NBP3-38198-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRODH Polyclonal antibody specifically detects PRODH in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| PRODH | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS (pH 7.3), 50% glycerol | |
| 5625 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 1.5.99.8, FLJ33744, HSPOX2, mitochondrial, p53-induced gene 6 protein, PIG6MGC148078, POX2, PRODH2MGC148079, proline dehydrogenase (oxidase) 1, proline dehydrogenase (proline oxidase ), Proline oxidase, proline oxidase, mitochondrial, SCZD4, tumor protein p53 inducible protein 6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 321-600 of human PRODH (NP_057419.4).,, Sequence:, IDSRTKLSKHLVVPNAQTGQLEPLLSRFTEEEELQMTRMLQRMDVLAKKATEMGVRLMVDAEQTYFQPAISRLTLEMQRKFNVEKPLIFNTYQCYLKDAYDNVTLDVELARREGWCFGAKLVRGAYLAQERARAAEIGYEDPINPTYEATNAMYHRCLDYVLEELKHNAKAKVMVASHNEDTVRFALRRMEELGLHPADHQVYFGQLLGMCDQISFPLGQAGYPVYKYVPYGPVMEVLPYLSRRALENSSLMKGTHRERQLLWLELLRRLRTGNLFHRPA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title