missing translation for 'onlineSavingsMsg'
Learn More

PRODH Antibody (3A9), Novus Biologicals™

Product Code. 18329378 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329378 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18329378 Supplier Novus Biologicals Supplier No. H00005625M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PRODH Monoclonal antibody specifically detects PRODH in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRODH
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3A9
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_057419
Gene Alias EC 1.5.99.8, FLJ33744, HSPOX2, mitochondrial, p53-induced gene 6 protein, PIG6MGC148078, POX2, PRODH2MGC148079, proline dehydrogenase (oxidase) 1, proline dehydrogenase (proline oxidase ), Proline oxidase, proline oxidase, mitochondrial, SCZD4, tumor protein p53 inducible protein 6
Host Species Mouse
Immunogen PRODH (NP_057419, 441 a.a. ~ 540 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LVRGAYLAQERARAAEIGYEDPINPTYEATNAMYHRCLDYVLEELKHNAKAKVMVASHNEDTVRFALRRMEELGLHPADHRVYFGQLLGMCDQISFPLGQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 5625
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.