missing translation for 'onlineSavingsMsg'
Learn More

Prochlorperazine Sulfoxide, TRC

Product Code. 30411288
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411288 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411288 Supplier Toronto Research Chemicals Supplier No. TRCP7558101MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prochlorperazine Sulfoxide
CAS 10078-27-0
Molecular Formula C20 H24 Cl N3 O S
Quantity 1 mg
Synonym 2-Chloro-10-[3-(4-methyl-1-piperazinyl)propyl]-10H-phenothiazine 5-oxide, 2-Chloro-10-[3-(4-methyl-1-piperazinyl)propyl]phenothiazine 5-oxide, Meterazine sulfoxide, Prochlorperazine sulfoxide
InChI Formula InChI=1S/C20H24ClN3OS/c1-22-11-13-23(14-12-22)9-4-10-24-17-5-2-3-6-19(17)26(25)20-8-7-16(21)15-18(20)24/h2-3,5-8,15H,4,9-14H2,1H3
SMILES CN1CCN(CCCN2c3ccccc3S(=O)c4ccc(Cl)cc24)CC1
IUPAC Name 2-chloro-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine 5-oxide
Molecular Weight (g/mol) 389.94
Formula Weight 389.1329
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.