missing translation for 'onlineSavingsMsg'
Learn More

Prochlorperazine Dimaleate, TRC

Product Code. 18091050
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18091050 100 mg 100mg
18091820 5 mg 5mg
18020620 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18091050 Supplier Toronto Research Chemicals Supplier No. TRCP755805100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-02-6
Molecular Formula C20 H24 Cl N3 S . 2 C4 H4 O4
Molecular Weight (g/mol) 606.09
Synonym Prochlorperazine maleate, 2-Chloro-10-[3-(4-methylpiperazin-1-yl)propyl]-10H-phenothiazine bis[hydrogen (Z)-butenedioate], 10H-Phenothiazine, 2-chloro-10-[3-(4-methyl-1-piperazinyl)propyl]-, (2Z)-2-butenedioate (1:2), 10H-Phenothiazine, 2-chloro-10-[3-(4-methyl-1-piperazinyl)propyl]-, (Z)-2-butenedioate (1:2), Phenothiazine, 2-chloro-10-[3-(4-methyl-1-piperazinyl)propyl]-, dimaleate (6CI), Phenothiazine, 2-chloro-10-[3-(4-methyl-1-piperazinyl)propyl]-, maleate (1:2) (8CI), 2-Chloro-10-[3-(4-methyl-1-piperazinyl)propyl]phenothiazine dimaleate, Buccastem, Compazine, Emetiral, Meterazin maleate, Pasotomin, Prochloroperazine dimaleate, Prochlorperazine dimaleate, Prochlorperazine hydrogen maleate, Prochlorperazine maleate, Stemetil, Stemetil dimaleate, Vertigon
IUPAC Name (Z)-but-2-enedioic acid;2-chloro-10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine
SMILES CN1CCN(CCCN2c3ccccc3Sc4ccc(Cl)cc24)CC1.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H24ClN3S.2C4H4O4/c1-22-11-13-23(14-12-22)9-4-10-24-17-5-2-3-6-19(17)25-20-8-7-16(21)15-18(20)24;2*5-3(6)1-2-4(7)8/h2-3,5-8,15H,4,9-14H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
Formula Weight 605.1599
Percent Purity >95%
Chemical Name or Material Prochlorperazine Maleate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.