missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SI Analytics™ ProcessLine Metal Combination Electrode
Shop All SI Analytics Products
Click to view available options
Connector Types:
Screw Plug Head
Unit Size:
Each
Description
- Low-maintenance
- Metal combination electrode
- Glass shaft
- Silamid™ reference
- 2 hole diaphragm
- DuraLid reference system
- Sensor platinum disk
- Screw plug head
Specifications
Specifications
| Connector Types | Screw Plug Head |
| Material | Glass Shaft |
| pH Range | 0 to 14 |
| Temperature (Metric) Range | 0°C to 130°C |
| Length (Metric) | 120 mm |
| Diameter (Metric) | 12 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction