missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Memmert™ Process-Dependent Electromagnetic Door Lock for UN55plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN55plus Oven
Unit Size:
Each
Description
- Accessory
- Door lock
- For units with TwinDISPLAY only
Specifications
Specifications
| For Use With (Equipment) | UN55plus Oven |
| Type | Electromagnetic Door Lock |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction