Learn More
Memmert™ Process-Dependent Electromagnetic Door Lock For Un160Pa
Shop All Memmert Products
Click to view available options
Type:
Electromagnetic Door Lock
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Models U, UF TS, UNpa, S, I, ICP, IPP, IPS, HPP, ICH |
| Type | Electromagnetic Door Lock |