Learn More
Invitrogen™ Universal Assay Buffer (1X)
Description
The ProcartaPlex Universal Assay Buffer (1X) is developed for use with the ProcartaPlex assays. The ProcartaPlex Universal Assay Buffer (1X) is used for dilution of serum and plasma samples.
The ProcartaPlex Universal Assay Buffer (1X) is included in our basic kits and off the shelf panels. Additionally, we offer the Universal Assay Buffer as a stand-alone item.
Reported Application
Multiplex Immunoassay
Specifications
Specifications
| Product Type | Buffer |
| Quantity | 1 each |
| Species | Canine, Mouse, Rat, Porcine, Non-Human Primate, Human |
| For Use With (Equipment) | Luminex™ Instruments |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.