missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Rat Antibody Isotyping Panel (6 plex)

Product Code. 15562457
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 15562457

Brand: Invitrogen™ EPX06030123901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • Rat Isotyping Standard Mix
  • Rat Isotyping Bead Mix
  • Rat Isotyping Detection Antibody Mix
  • Reading Buffer
  • Wash Buffer 10x
  • Universal Assay Buffer 10x
  • Detection Antibody Diluent
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • Our assays are designed for use with the Luminex MAGPIX™, Luminex 100, Luminex 200, or Bio-Rad Bio-Plex analyzers
  • This kit does not contain a Multiscreen Filter Plate
  • It is recommended to dilute samples when measured in serum or plasma
  • The recommended sample dilution factor is 1:40,000
TRUSTED_SUSTAINABILITY

Specifications

Quantity 96 Tests
Species Rat
For Use With (Application) Multiplex Immunoassay
Protein Subtype IgA, IgG1, IgG2a, IgG2b, IgG2c, IgM
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.