missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-6 Mouse ProcartaPlex™ Simplex Kit, High Sensitivity

Product Code. 15562557
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Product Code. Quantity unitSize
15562557 96 Tests 96 tests
1 options
This item is not returnable. View return policy

Product Code. 15562557

Brand: Invitrogen™ EPXS01020603901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • concentrated Magnetic Beads coated with monoclonal antibody to mouse IL-6
  • mouse IL-6 Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-mouse IL-6 monoclonal antibody

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

When combining multiple Simplex kits (i.e., when not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

About ProcartaPlex Assays for the Luminex Platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.
More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g. Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

TRUSTED_SUSTAINABILITY

Specifications

Product Type High Sensitivity Simplex Kit
Quantity 96 Tests
Species Mouse
Combinability Combinable (High Sensitivity Only)
Bead Type IL-6 [28]
Assay Sensitivity See Certificate of Analysis
Assay Range As determined for Lot 1: 0.70 - 2,850 pg/mL
Format High Sensitivity Simplex Kit
For Use With (Equipment) Luminex™ Instruments
Gene Interleukin 6
Gene Alias IL-6
Gene ID (Entrez) 16193
Gene Symbol IL6
Product Line ProcartaPlex
Protein Interleukin-6
Protein Subtype IL-6, B-cell hybridoma growth factor, Interleukin HP-1
Sample Type Serum, Plasma, Cell Culture Supernatants
Sample Volume Serum, Plasma: 25 μL, CCS: 50 μL
UniProt ID P08505
Content And Storage

• 1 vial Capture Beads (50X)
• 1 vial biotinylated Detection Antibody (50X)
• 2 vials Mouse Standard Mix A - HS (lyophilized)

Store at 2°C to 8°C.

Shipping Condition Wet Ice
Research Area Immunology, Oncology, Neurobiology, Toxicology, Cardiovascular
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.