missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Mouse Cytokine Panel 1B (10 plex)

Product Code. 15502477
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15502477 96 Tests 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15502477 Supplier Invitrogen™ Supplier No. EPX10026091901
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

  • Mouse Cytokine 1B Panel Standard Mix
  • Mouse Cytokine 1B Panel Bead Mix
  • Mouse Cytokine 1B Panel Detection Antibody Mix
  • Reading Buffer
  • Wash Buffer 10x
  • Streptavidin-PE
  • Universal Assay Buffer
  • Detection Antibody Diluent
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • This kit does not contain a Multiscreen Filter Plate
TRUSTED_SUSTAINABILITY

Specifications

Quantity 96 Tests
Species Mouse
For Use With (Application) Multiplex Immunoassay
Protein Subtype ENA-78, G-CSF, IFN alpha, IL-1 alpha, IL-15/IL-15R complex, IL-28, IL-3, IL-31, LIF, M-CSF
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.