missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Mouse Antibody Isotyping Panel 2, 7plex

Product Code. 15522467
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy
Certain inbred strains of mice such as C57Bl/6, C57Bl/10, SJL, and NOD with the Igh1-b allele do not have the gene for IgG2a, but instead express IgG2c. Therefore, Thermofisher Scientific included IgG2c into the ProcartaPlex Mouse Isotyping Panel 2.

Product Code. 15522467

Brand: Invitrogen™ EPX07020816901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Certain inbred strains of mice such as C57Bl/6, C57Bl/10, SJL, and NOD with the Igh1-b allele do not have the gene for IgG2a, but instead express IgG2c. Therefore, Thermofisher Scientific included IgG2c into the ProcartaPlex Mouse Isotyping Panel 2.

Includes

Mouse Isotyping Standard Mix, Mouse IgG2c Standard, Mouse Isotyping Bead Mix, Mouse Isotyping Detection Antibody Mix, Reading Buffer, Wash Buffer 10X, Universal Assay Buffer, Universal Assay Buffer 10X, Detection Antibody Diluent, PCR 8-Tube Strip, Adhesive Film, Greiner Microplate (black, 96-well)

The ProcartaPlex Mouse Antibody Isotyping Panel 2 7plex enables determination of antibody heavy chain subclasses by analyzing 7 protein targets using Luminex xMAP technology.

The ProcartaPlex Mouse Antibody Isotyping Panel 2 7plex enables determination of antibody heavy chain subclasses by analyzing 7 protein targets using Luminex xMAP technology. Antibody isotyping is a critical and beneficial tool in hybridoma development. Simultaneous detection of these targets can help researchers explore analysis of immunoglobulin isotypes during and after hybridoma development and to assess immune responses. The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup. It is not combinable with simplexes or other panels.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit our ProcartaPlex Immunoassays page for more information and available products.

Target list [bead region]: IgA [38], IgE [42], IgG1 [12], IgG2b [21], IgG2c [43], IgG3 [25], IgM [52]

Recommended sample dilution: Serum, Plasma: 1:40,000; Cell culture supernatant: 1:200

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

Features include:

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 96 Tests
Species Mouse
Combinability Non-Combinable
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
Format Multiplex Kit
For Use With (Equipment) Luminex™ Instruments
Product Line ProcartaPlex™
Sample Type Serum, Plasma, Cell Culture Supernatants
Sample Volume Serum, Plasma: 25 μL, CCS: 50 μL
Content And Storage

• 2 vials Mouse Isotyping Standard Mix (lyophilized)
• 2 vials Mouse IgG2c Standard (lyophilized)
• 1 vial Capture Bead Mix (1X)
• 1 vial Biotinylated Detection Antibody Mix (50X)
• 1 bottle Reading Buffer (1X)
• 1 bottle Wash Buffer (10X)
• 1 bottle Universal Assay Buffer (10X)
• 1 bottle Detection Antibody Diluent (1X)
• 8-tube strip
• Adhesive film
• Flat-bottom 96-well plate, black

Store at 2°C to 8°C

Shipping Condition Wet Ice
Research Area Antibody Isotyping
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.