missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ ProcartaPlex Human PD-L1 (CD274) Simplex
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
Includes
- concentrated Magnetic Beads coated with monoclonal antibody to human PD-L1 (CD274)
- human PD-L1 (CD274) Standard (lyophilized)
- Biotin-Conjugate (50x) anti-human PD-L1 (CD274) monoclonal antibody
- Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
- ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
- PD-L1 cannot be combined with IL-4 and IL-1RA in plasma/serum samples
Specifications
Specifications
| Quantity | 96 Tests |
| Species | Human |
| For Use With (Application) | Multiplex Immunoassay |
| Sample Volume | Serum, Plasma: 25 μL; CCS: 50 μL |
| Synonym | B7H1, PD-L1, PDL-1, PDL1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction