missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Human Neuroscience Panel, 21plex

Product Code. 31905078
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31905078 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31905078 Supplier Invitrogen™ Supplier No. EPX21015839901
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The ProcartaPlex Human Neuroscience Panel, 21plex, is the biggest ProcartaPlex panel for neurological research and enables the quantification of 21 protein targets in a single well using Luminex xMAP technology.

The ProcartaPlex Human Neuroscience Panel, 21plex, is the biggest ProcartaPlex panel for neurological research and enables the quantification of 21 protein targets in a single well using Luminex xMAP technology. The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup. It is not combinable with simplexes or other panels.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

Target list [bead region]: amyloid beta 1-42 [42], alpha-synuclein (total) [53], BDNF [57], CNTF [36], FGF-21 [76], GDNF [44], GFAP [22], kallikrein-6 (KLK6) [14], NCAM-1 [48], neurogranin (NRGN) [75], NF-H [33], NF-L [78], NGF beta [55], NSE [12] S100B [18], TAU (total) [46], TAU [p181] [61], TDP-43 [74], TREM-2 [13] UCHL-1 [19], and YKL-40 (CHI3L1) [34]

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum or 50 μL of cell culture supernatant and just four hours to obtain analyzed results.

Features include:

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, xMAP INTELLIFLEX, xMAP INTELLIFLEX DR-SE, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 96 Tests
Species Human
Combinability Non-Combinable
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
For Use With (Equipment) Luminex™ Instruments
Interassay CV Less than 15%
Intraassay CV Less than 15%
Multiplex Size Plex No. 21 plex
Product Line ProcartaPlex
Sample Type Serum, Plasma, Cerebrospinal Fluid
Sample Volume Serum, Plasma, CSF: 25 μL
Shipping Condition Wet Ice
Research Area Neurobiology
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.