missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ ProcartaPlex Human Eotaxin-2 (CCL24) Simplex

Product Code. 15522257
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 15522257

Brand: Invitrogen™ EPX01A12150901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

  • concentrated Magnetic Beads coated with monoclonal antibody to human Eotaxin-2
  • human Eotaxin-2 Standard (lyophilized)
  • Biotin-Conjugate (50x) anti-human Eotaxin-2 monoclonal antibody

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • Quantitate multiple targets in cell/tissue culture supernatants, serum, and plasma
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
TRUSTED_SUSTAINABILITY

Specifications

Quantity 96 Tests
Species Human
For Use With (Application) Multiplex Immunoassay
Sample Type Cell Culture Supernatant, Plasma, Serum
Sample Volume Serum, Plasma: 25 μL; CCS: 50 μL
Synonym CCL24, myeloid progenitor inhibitory factor 2, eosinophil chemotactic protein 2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.