missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Detection Antibody Diluent
Click to view available options
Quantity:
1 each
Unit Size:
Each
Specifications
Specifications
| Product Type | Diluent |
| Quantity | 1 each |
| Species | Human |
| For Use With (Equipment) | Luminex™ Instruments |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction