missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Coagulation 3-Plex Human ProcartaPlex™ Panel 2

Product Code. 15532447
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 15532447

Brand: Invitrogen™ EPX03010823901

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • Human Coagulation Panel 2 Reference Plasma Standard
  • Human Coagulaton Panel 2 Bead Mix
  • Human Coagulation Panel 2 Detection Antibody Mix
  • Reading Buffer
  • Wash Buffer 10x
  • Streptavidin-PE
  • Universal Assay Buffer 10X
  • Detection Antibody Diluent
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

  • The human Coagulation panels are designed for the use with plasma samples (EDTA, Citrate)
  • ProcartaPlex assays use Luminex™ xMAP technology for the multi-analyte detection of secreted proteins including cytokines, chemokines, growth factors and other proteins
  • This kit does not contain a Multiscreen Filter Plate
TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 96 Tests
Species Human
Assay Sensitivity Less than 15%
Assay Range See Certificate of Analysis
Format Multiplex Kit
For Use With (Equipment) Luminex Instruments
Product Line ProcartaPlex
Sample Type Plasma (EDTA, Citrate)
Sample Volume Plasma: 25 μL
Content And Storage
  • 2 vials Human Reference Plasma Standard
  • 1 vial Capture Bead Mix (1X)
  • 1 vial Biotinylated Detection Antibody Mix (50X)
  • 1 bottle Detection Antibody Diluent (1X)
  • 1 bottle Reading Buffer (1X)
  • 1 bottle Streptavidin-PE (1X)
  • 1 bottle Universal Assay Buffer (10x)
  • 1 bottle Wash Buffer (10x)
  • 8-tube strip
  • Adhesive film
  • Flat-bottom 96-well plate, black
  • Store at 2°C to 8°C
Shipping Condition Wet Ice
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.