missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Coagulation 6-Plex Human ProcartaPlex™ Panel 1

Product Code. 15532457
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 15532457

Brand: Invitrogen™ EPX06010824901

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • Human Coagulation Panel 1 Reference Plasma Standard
  • Human Coagulaton Panel 1 Bead Mix
  • Human Coagulation Panel 1 Detection Antibody Mix
  • Reading Buffer
  • Wash Buffer 10x
  • Streptavidin-PE
  • Universal Assay Buffer 10X
  • Detection Antibody Diluent
  • PCR 8-Tube Strip
  • Adhesive Film
  • Greiner Microplate black 96 well

Ideal solution for assessing multiple protein biomarkers in a single sample. Explore our broad portfolio of pre-configurated panels and combinable single analytes.

The Human Coagulation 6-Plex ProcartaPlex Panel 1 enables research of wound healing and clotting disorders by analyzing 6 protein targets in a single well using Luminex xMAP technology. Simultaneous detection of these targets can help explore diseases such as hemophilia or von Willebrand disease. The panel is provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup. It is not combinable with simplexes or other panels. It complements the Coagulation 6-Plex ProcartaPlex Panel 3 (Cat. No. EPX040-10825-901), Coagulation 3-Plex Human ProcartaPlex Panel 2 (Cat. No. EPX030-10823-901), Factor X Simplex Kit (Cat. No.EPX01A-12152-901), and Fibrinogen Simplex Kit (Cat. No. EPX01A-12153-901) that provides 9 additional protein targets in the same research area.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

Target list [bead region]: Antithrombin [27], CRP [15], Factor XI [29], Factor XII [30], Factor XIII [47], Prothrombin [34]

Recommended sample pre-dilution: Plasma (Citrate, EDTA): 1:5000

About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

Features include:
• Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
• More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
• Well-established Luminex technology highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25–50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

The Luminex beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create spectrally unique signatures that can be identified by the Luminex xMAP detection systems (e.g., Luminex 200, FLEXMAP 3D, and MAGPIX). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a multiplexed assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 96 Tests
Species Human
Combinability Non-Combinable
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
Format Multiplex Kit
For Use With (Equipment) Luminex™ Instruments
Product Line ProcartaPlex
Sample Type Plasma (EDTA, Citrate)
Sample Volume Plasma: 25 μL
Content And Storage

• 2 vials Human Reference Plasma Standard
• 1 vial Capture Bead Mix (1X)
• 1 vial Biotinylated Detection Antibody Mix (50X)
• 1 bottle Reading Buffer (1X)
• 1 bottle Wash Buffer (10X)
• 1 bottle Streptavidin-PE (1X)
• 1 bottle Universal Assay Buffer (10X)
• 1 bottle Detection Antibody Diluent (1X)
• 8-tube strip
• Adhesive film
• Flat-bottom 96-well plate, black

Store at 2°C to 8°C

Shipping Condition Wet Ice
Research Area Coagulation
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.