Learn More
Invitrogen™ ProcartaPlex™ Flat Bottom Plate, 96-well
Product Code. 18739279
Click to view available options
Quantity:
96 tests
Unit Size:
Each
Product Code. 18739279
Brand: Invitrogen™ EPX88182000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This black ProcartaPlex 96-well flat-bottom plate is ideal for use with all ProcartaPlex Luminex protein assays.
This black ProcartaPlex 96-well flat-bottom plate is ideal for use with all ProcartaPlex Luminex protein assays.
This plate is included in all ProcartaPlex multiplex panel kits, species-specific basic kits, and as part of custom mix and match panels. For bulk orders, please contact your sales representative.
Note that the reusable lid (Cat. No. EPX-88104-000) is sold separately.
About ProcartaPlex assays for the Luminex platform
ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to detect and quantitate up to 80 protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.
ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, non-human primate, porcine, and canine). Visit thermofisher.com/procartaplex for more information and available products.
Related products
  • ProcartaPlex Microplate Lid (Cat. No. EPX-88104-000)
TRUSTED_SUSTAINABILITY
Specifications
Lid Without Lid
Product Line ProcartaPlex
Color Black
Disposable Single-use
Product Type Plate
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Hydrophobic
Well Shape Flat Bottom (F-well)
Quantity 96 tests
Fit for Purpose Certified Quality