missing translation for 'onlineSavingsMsg'
Learn More

Miele Professional™ ProCare Lab 11 MA

Product Code. 15410425 Shop All Miele Products
Change view
Click to view available options
Volume (Metric):
10 kg
Unit Size:
10kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
15410425 10 kg 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15410425 Supplier Miele Professional™ Supplier No. 10266720
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Powder cleaning agent, mildly alkaline, 10 kg. For optimum reprocessing of laboratory utensils.

  • Foam damping effect, free from oxidants
  • Suitable for the reprocessing phases in washer-disinfectors
  • Suitable for all types of materials
  • pH value 11.7 to 11.9
TRUSTED_SUSTAINABILITY

Specifications

pH 11.7 to 11.9
Description Powder cleaning agent
For Use With (Equipment) Suitable for the reprocessing phases in washer-disinfectors
Volume (Metric) 10 kg
compliance-icons compliance-icons
Product Identifier
  • miele procare lab 11 MA
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: ethoxylated fatty alcohol, disodium metasilicate pentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.